Anti-GABRA1 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-D43
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, alpha 1
- Antigen symbol:GABRA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:29705
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-GABA(A) Receptor alpha 1 Antibody was produced in rabbit by repeated immunizations with synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH.
- Purification:affinity-purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-GABA(A) Receptor alpha 1 (Rabbit) Antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 51 kDa in size corresponding to the alpha 1 subunit of the GABA(A) Receptor.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: GABRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat