Anti-GABRD Mouse Monoclonal Antibody (Unconjugated) [clone: S151-3]
Catalog # ROCK200-301-F33
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, delta
- Antigen symbol:GABRD
- Clonality:Monoclonal
- Clone:S151-3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:Internal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:29689
- Antigen synonyms:GABAA-RD
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:GABA-A receptor-delta Antibody was produced in mice by repeated immunizations with a synthetic peptide corresponding to an internal region of rat GABA-A-R-Delta.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-GABA-A receptor delta Antibody is suitable for use in WB and IF microscopy. Expect a band approximately ~55kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:5000-1:25000; Immunofluorescence: 1.0-10µg/ml; Western Blot: 1-10µg/ml; Immunochemistry: 0.1-1.0µg/ml
Type: Primary
Antigen: GABRD
Clonality: Monoclonal
Clone: S151-3
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat