Anti-GABBR2 Rabbit Polyclonal Antibody
Catalog # ROCK612-401-D53
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) B receptor 2
- Antigen symbol:GABBR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Rat
- Western blot:Yes
- ChIP:Yes
- Gene ID:83633
- Antigen synonyms:Gpr51
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-GABA(B) Receptor 2 pS783 (Rabbit) antibody is suitable for use in Western Blotting and IF. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 102 kDa in size corresponding to GABA(B) receptor 2 protein.
Recommended Dilutions: Immunofluorescence: 1:500; Western Blot: 1:1000
Type: Primary
Antigen: GABBR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat