Anti-GABRA4 Rabbit Polyclonal Antibody
Catalog # ROCK612-401-D47
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, alpha 4
- Antigen symbol:GABRA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:140675
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-GABA(A) Receptor alpha 4 Antibody was produced by repeated immunizations with recombinant fusion Proteins from the cytoplasmic loop of the alpha 4 subunit.
- Purification:affinity-purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-GABA(A) Receptor alpha 4 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 64 kDa in size corresponding to the alpha 4 subunit of the GABA.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: GABRA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat