ContactUsLinkComponent
Attachable screw thread tubes
Attachable screw thread tubes
Catalog # WITG0.182.315
Supplier:  witeg Labortechnik
undefined
Attachable screw thread tubes
Catalog # WITG0.182.315
Supplier:  witeg Labortechnik
Specifications
  • Thread:
    SVL 15
  • Tube ext.Ø:
    16 mm
  • Total length:
    110 mm
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
DIN screw thread SVL.