Orders LinkContactUsLinkComponent
Sample vials, with cap
Sample vials, with cap
   Sustainable Options Available
Supplier:   EP SCIENTIFIC
Sample vials, with cap
Supplier:  EP SCIENTIFIC
   Sustainable Options Available
Certificates
Sample vials, with cap
5 Options Available Below
Product Details & Documents
Clear or amber sample vials made from borosilicate glass.
  • Designed for water sample collection for volatile organic analysis (VOA)
  • Optimised for use with automation
  • With open-top white PP cap bonded with 1,52 mm thin PTFE-lined silicone septum
  • Septum permanently bonded to the cap for secure sealing
  • Vial shoulder slope optimized to prevent air bubble entrapment
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis but without certificate. Processed vials are processed in the same manner as certified vials and come with all the features of certified vials, but vials are not barcoded and CofA is not included.
Specifications for Product Family
Specifications
  • Catalog No:
  • EPSCT136-0040
  • EPSCT236-0040
  • EPSCT336-0040
  • EPSCT246-0040
  • EPSCT346-0040
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Type
    Capacity
    Ø×H
    Colour
    Cap included
    Cap type
    Cap Size
    Septum
    Thickness
    Availability
    Price
  • EPSCT136-0040
    Unprocessed
    Unprocessed
    Unprocessed, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Clear
    Yes
    Screw cap, open-top cap
    24-414
    Bonded
    1,52 mm
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 337,00 €
    337,00 €
    1 * 72 ST
       337,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCT136-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCT236-0040
    VOA processed
    VOA processed
    VOA processed, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Clear
    Yes
    Screw cap, open-top cap
    24-414
    Bonded
    1,52 mm
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 485,00 €
    485,00 €
    1 * 72 ST
       485,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCT236-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCT336-0040
    VOA certified
    VOA certified
    VOA certified, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Clear
    Yes
    Screw cap, open-top cap
    24-414
    Bonded
    1,52 mm
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 587,00 €
    587,00 €
    1 * 72 ST
       587,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCT336-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCT246-0040
    VOA processed
    VOA processed
    VOA processed, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Amber
    Yes
    Screw cap, open-top cap
    24-414
    Bonded
    1,52 mm
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 485,00 €
    485,00 €
    1 * 72 ST
       485,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCT246-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCT346-0040
    VOA certified
    VOA certified
    VOA certified, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Amber
    Yes
    Screw cap, open-top cap
    24-414
    Bonded
    1,52 mm
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 587,00 €
    587,00 €
    1 * 72 ST
       587,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCT346-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact