Order Entry
Austria
ContactUsLinkComponent
Anti-HSP16.9 Rabbit Polyclonal Antibody
Anti-HSP16.9 Rabbit Polyclonal Antibody
Catalog # AGRIAS122570
Supplier:  Agrisera
Anti-HSP16.9 Rabbit Polyclonal Antibody
Catalog # AGRIAS122570
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HSP16.9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Zea mays
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Molecular weight:
    16.9 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    recombinant protein of Triticum aestivum Q41560
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: HSP16.9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Zea mays