Anti-Acd1 Rabbit Polyclonal Antibody
Catalog # AGRIAS111783
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:Accelerated Cell Death 1
- Antigen symbol:ADC-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:PaO,áLls1
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant PaO from Arabidopsis thaliana Q9FYC2, At3g44880
- Size:100 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ADC-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana