Anti-Beta-CA1/ beta-CA2 Rabbit Polyclonal Antibody
Catalog # AGRIAS111737
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:beta CA1/2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant Chlamydomonas reinhardtii mitochondrial CA, as described in Villand et al. 1997. Accession number Q39590 and Q39589
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: beta CA1/2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana