Anti-Sml1 Rabbit Polyclonal Antibody
Catalog # AGRIAS10847
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:SML1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Synechocystis sp. pcc 6803
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from known S.cerevisie Sml1 sequence. Gene ID: 854945
- Size:200 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SML1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Synechocystis sp. pcc 6803