Order Entry
Austria
ContactUsLinkComponent
Anti-IgG (H+L) Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgG (H+L) Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # AGRIAS101537
Supplier:  Agrisera
Anti-IgG (H+L) Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # AGRIAS101537
Supplier:  Agrisera

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Sheep
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rabbit
  • Cross adsorption:
    No
  • Storage temperature:
    +2°C to +8 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Purified rabbit IgG (H&L)
  • Purification:
    purified
  • Size:
    2 mg
  • Pk:
    2 mg

Specifications

About this item

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Sheep
Isotype:
Reactivity: Rabbit