Anti-AKINB1 Rabbit Polyclonal Antibody
AGRIAS09460
: Agrisera
- Antibody type:Primary
- Antigen symbol:AKINB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated peptide derived from Arabidopsis thaliana AKIN beta-1 Q84VQ1, At5g21170
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: AKINB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human