Anti-14-3-3C Rabbit Polyclonal Antibody
Catalog # AGRIAS08317
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:C. reinhardtii,N. tabacum
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:synthetic peptide specific to the barley C isoform, conjugated to KLH
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: 14-3-3 C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii, N. tabacum