Anti-AtpA Rabbit Polyclonal Antibody
Catalog # AGRIAS08304
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:ATP synthase CF1 alpha chain
- Antigen symbol:ATPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Spinacia oleracea,C. reinhardtii,A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant maize chloroplast AtpA P05022
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ATPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii, Spinacia oleracea