Anti-HSP21 Rabbit Polyclonal Antibody
AGRIAS08285
: Agrisera
- Antibody type:Primary
- Antigen symbol:HSP21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Cucumis sativus,Agave tequiliana var. weber,A. thaliana,Pinellia ternata,Silene vulgaris
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant Hsp21 protein derived from the sequence of Arabidopsis thaliana hsp21 P31170, At4g27670
- Size:100 µl
- Pk:50 µl
Type: Primary
Antigen: HSP21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Agave tequiliana var. Weber, A. thaliana, Cucumis sativus, Pinellia ternata, Silene vulgaris