Anti-DsRed-Monomer IgG1 Monoclonal Antibody (Magnetic beads) [clone: OTI4C7 (formerly 4C7)]
Catalog # ORIGTA183039
Supplier: OriGene
Specifications
- Antibody type:Tag
- Antigen symbol:DsRed-Monomer
- Clonality:Monoclonal
- Clone:OTI4C7 (formerly 4C7)
- Conjugation:Magnetic beads
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Form:Concentrated
- Storage buffer:PBS, pH 7,4, containing 0,1% BSA, 0,02% NaN3 and 50% glycerol with final concentration 5mg/mL beads
- Molecular weight:25.4 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This antibody is raised against a recombinant protein of DsRed-Monomer expressed in HEK293 cells.
- Purification:affinity chromatography
- Pk:1 ml
Specifications
About this item
Type: Tag
Antigen: DsRed-Monomer
Clonality: Monoclonal
Clone: OTI4C7 (formerly 4C7)
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: