Anti-IFNGR2 Mouse Polyclonal Antibody [clone: 4S.B3]
Catalog # ORIGTA320354
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Interferon gamma receptor 2
- Antigen symbol:IFNGR2
- Clonality:Polyclonal
- Clone:4S.B3
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: IFNGR2
Clonality: Polyclonal
Clone: 4S.B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human