Anti-HLA-G Mouse Polyclonal Antibody [clone: 87G]
ORIGTA320347
: OriGene
- Antibody type:Primary
- Antigen symbol:HLA-G
- Clonality:Polyclonal
- Clone:87G
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: HLA-G
Clonality: Polyclonal
Clone: 87G
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human