Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 3E10]
Catalog # ORIGCF803247
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Carcinoembryonic antigen-related cell adhesion molecule 5
- Antigen symbol:CEACAM5
- Clonality:Monoclonal
- Clone:3E10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:35 to 680
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 35-680 of human CEACAM5 (NP_004354) produced in SF9 cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human