Order Entry
Austria
ContactUsLinkComponent
Anti-KI67 Rat Monoclonal Antibody [clone: 3B6]
Anti-KI67 Rat Monoclonal Antibody [clone: 3B6]
  ORIGCF801343
 :  OriGene
Anti-KI67 Rat Monoclonal Antibody [clone: 3B6]
  ORIGCF801343
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    KI67
  • Clonality:
    Monoclonal
  • Clone:
    3B6
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    1160 to 1493
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 1160-1493 of human MKI67 (NP_002408) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human