Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 3H2]
Catalog # ORIGCF800059
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:UBE2E3
- Clonality:Monoclonal
- Clone:3H2
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:22.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human UBE2E3 (NP_006348) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000
Type: Primary
Antigen: UBE2E3
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human