Anti-RIPK3 Mouse Monoclonal Antibody [clone: 2H8]
Catalog # ORIGCF803099
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Receptor-interacting serine-threonine kinase 3
- Antigen symbol:RIPK3
- Clonality:Monoclonal
- Clone:2H8
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:233 to 518
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 233-518 of human RIPK3 (NP_006862) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: RIPK3
Clonality: Monoclonal
Clone: 2H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human