Orders LinkContactUsLinkComponent
Vacuum controller
Vacuum controller
Vacuum controller, CVC 3000 E
Catalog # VACU20683180
Supplier:   VACUUBRAND
Vacuum controller
Catalog # VACU20683180
Supplier:  VACUUBRAND
Vacuum controller
Vacuum controller
Vacuum controller, CVC 3000 E
Price
2 520,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The VACUU.LAN local vacuum network connects several workstations to one vacuum pump. The various modules make it possible to connect almost all popular vacuum applications such as rotary evaporators, vacuum concentrators, gel dryers, drying ovens and filter systems. It can be integrated into new laboratory furniture at the time of assembly or subsequently installed in existing laboratories.
  • Individual connection configuration, vacuum connections to suit the requirements of the workstation
  • Integrated non-return valves to prevent cross-contamination, even with a low pressure differential
  • Demand-driven vacuum generation, automatic switching in and out of the vacuum pump
  • Flexible design, easily adapted to changes in requirements or in the vacuum connection
  • Practical design, integrated installation (installation conduit) or wall-mounting
CVC 3000E
The modules are fully assembled or pre-assembled and ready for connection. Different connecting elements enable integrated installation into newly planned laboratory furniture, subsequent installation in existing laboratories or upgrading of existing older networks. Each VCL module consists of a connecting element and a functional element.
Dokumente
Specifications
  • Description:
    Vacuum controller, CVC 3000 E