Order Entry
Austria
ContactUsLinkComponent
n-Hexane ≥95%, PESTINORM® Supra Trace for organic trace analysis
Flammable
Health
Toxic
Environment
n-Hexane ≥95%, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85389.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Toxic
Environment
n-Hexane ≥95%, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85389.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 95%
  • Evaporation residue:
    Max 0.0003%
  • Colouration:
    Max 10 APHA
  • Water:
    Max 0.005%
  • GC/ECD Dioxins, Furans & PCB's:
    Max 2 ng/l
  • GC/ECD any pesticide (as Lindane):
    Max 2 ng/l
  • GC/NPD any pesticide (as Parathion):
    Max 5 ng/l
  • GC/ECD 1,2,4-TCB to daca-PCB (as Lindane):
    Max 2 pg/ml
  • GC/ECD DCM to 1,2,4-TCB (as TCM):
    Max 1 ng/l
  • GC/FID C10 to C40 (as n-Decane):
    Max 2 ng/l
  • GC/MSD C10 to C40 (as Decane; 30-600amu):
    Max 2 ng/l
  • Filtered through 0.2um, filled under inert gas:
    Conforms

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94,3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under inert gas.
 :GHS02
 :GHS08
 :GHS06
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520