Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
 
n-Heptane ≥99.0%, PESTINORM® Supra Trace for organic trace analysis
  85388.320
 :  
undefined
 
 
 
 
n-Heptane ≥99.0%, PESTINORM® Supra Trace for organic trace analysis
  85388.320
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay (GC on anhydrous substance):
    99.0-100%
  • Evaporation residue:
    Max 0.0003%
  • Colouration:
    Max 10 APHA
  • Water:
    Max 0.005%
  • GC/ECD Dioxins, Furans & PCB's:
    Max 2 ng/l
  • GC/ECD any pesticide (as Lindane):
    Max 2 ng/l
  • GC/NPD any pesticide (as Parathion):
    Max 5 ng/l
  • GC/ECD 1,2,4-TCB to daca-PCB (as Lindane):
    Max 2 pg/ml
  • GC/ECD DCM to 1,2,4-TCB (as TCM):
    Max 1 ng/l
  • GC/FID C10 to C40 (as n-Decane):
    Max 2 ng/l
  • GC/MSD C10 to C40 (as Decane; 30-600amu):
    Max 2 ng/l
  • Filtered through 0.2um, filled under inert gas:
    Conforms

 

 

 :
 :100.20 g/mol
 :13,04679
 :98.4 °C (1013 hPa)
 :–90,6 °C
 :0.6838 g/cm³ (20 °C)
 :–4 °C
 :Ambient
 :205-563-8
 :1206
 :3,II
 :Filtered through a 0.2 µm filter, packaged under inert gas.
 :GHS02
 :GHS08
 :GHS07
 :GHS09
 :142-82-5
 :H₃C(CH₂)₅CH₃
 :MFCD00009544