Jars, sampling, wide mouth, with screw cap
Supplier: EP SCIENTIFIC
Certificates
About this item
Jars, clear or amber Type III soda-lime glass, with PTFE-lined white polypropylene closed-top cap, designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analyses.
- Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals analyses
- Processed the same way as the certified jars but without certification documentation
- Unprocessed and ready for your own cleaning procedure
- Closures assembled on bottles
- Available as short or tall profile
Delivery: Certified jars supplied with Certificate of Analysis (CofA) in the case and barcoded with lot number for traceability.
Specifications
- Catalog No:
- EPSCV220-0125
- EPSC221-4000
- EPSC220-0060
- EPSC221-1000
- EPSC321-0250
- EPSC220-0125
- EPSC321-0500
- EPSC321-1000
- EPSC121-1000
- EPSC121-4000
- EPSC220-0250
- EPSC320-0250
- EPSC320-0500
- EPSC321-2000
- EPSC321-0125
- EPSCV221-1000
- EPSC221-0125
- EPSC120-0060
- EPSCV320-0125
- EPSC221-0250
- EPSC220-0500
- EPSC220-2000
- EPSC120-0125
- EPSC121-0125
- EPSC221-2000
- EPSC120-2000
- EPSC121-0250
- EPSCV320-0500
- EPSC120-0250
- EPSC320-0125
- EPSCV321-1000
- EPSC321-4000
- EPSC320-0060
- EPSC320-2000
- EPSC221-0500
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement