Order Entry
Austria
ContactUsLinkComponent
 
Tank, rectangular, PFA
  ENTEE92-01D-0215
 :  ENTEGRIS
undefined
Tank, rectangular, PFA
  ENTEE92-01D-0215
 :  ENTEGRIS

 

  • Capacity:
    23 L
  • Description:
    Process tank, rectangular, with 1/2" FNPT drain
  • W×D×H:
    406×203×279 mm
  • Pk:
    1

 

 

Injection molded rectangular tanks with dual capacity.

  • Available in single or dual capacity
  • Available with or without covers
  • Tanks should be properly supported in heated applications

Maximum recommended working temperature: 180 °C.

Mounting hole dimension (L×W): 419×216 mm.