Anti-IgG Mouse Antibody (Biotin)
Catalog # BSBTBM2001-0.5
Supplier: Boster Bio
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Biotin
- Host:Mouse
- Cross adsorption:No
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: