Anti-Steptavidin Antibody (Peroxidase)
Catalog # BSBTBA1088-0.5
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Steptavidin
- Conjugation:HRP (Horseradish Peroxidase)
- Cross adsorption:No
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type: Primary
Antigen: Steptavidin
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity: