Order Entry
Austria
ContactUsLinkComponent
Anti-Protein A Antibody (Peroxidase)
Anti-Protein A Antibody (Peroxidase)
Catalog # BSBTBA1080-0.5
Supplier:  Boster Bio
Anti-Protein A Antibody (Peroxidase)
Catalog # BSBTBA1080-0.5
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Protein A
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Cross adsorption:
    No
  • Size:
    500 μg
  • Pk:
    0,5 mg

Specifications

About this item

Type: Tag
Antigen: Protein A
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity: