Dropping pipettes
Supplier: SciLabware
Jump to:
About this item
Soda-lime glass.
- With integral ground glass cone on pipette
- Supplied with rubber teats
- Bottles available separately
3 Options Available Below
Product Details & Documents
Soda-lime glass.
- With integral ground glass cone on pipette
- Supplied with rubber teats
- Bottles available separately
Product Family Options
Product Information
- AvailabilityPrice
- BIBBSBD050Dropping pipettes with silicone teat, for 50 ml bottles, joint size 14/15Dropping pipettes with silicone teat, for 50 ml bottles, joint size 14/15Log in for availability1 * 5 ST85,60 €
- BIBBSBD100Dropping pipettes with silicone teat, for 100 ml bottles, joint size 14/23Dropping pipettes with silicone teat, for 100 ml bottles, joint size 14/23Log in for availability1 * 5 ST87,60 €
- BIBBSBD250Dropping pipettes with silicone teat, for 250 ml bottles, joint size 19/26Dropping pipettes with silicone teat, for 250 ml bottles, joint size 19/26Log in for availability1 * 5 ST49,80 €