Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Syringes
Syringes for Chromatography
Syringes for HPLC autosamplers
Syringes for HPLC autosamplers
Syringes for HPLC autosamplers
Catalog # HAMI81260
Supplier:
HAMILTON BONADUZ
Expand image
Syringes for HPLC autosamplers
Catalog # HAMI81260
Supplier:
HAMILTON BONADUZ
Specifications
Type:
Model 1750 C
Volume:
500 µl
Description:
Syringes, 1750 C, 1/4-28 thread
Pk:
1
Specifications
About this item
Product Details
Product Details