Tubings, insulated
Supplier: LAUDA
Certificates
About this item
Metal hoses with insulation for connections with thread M16×1.
- Temperature resistance: −90 to +150 °C.
Frequently Bought Together
Metal hoses with insulation for connections with thread M16×1.