Order Entry
Austria
ContactUsLinkComponent
Twaddle hydrometer
Twaddle hydrometer
Catalog # GERI0549S
Supplier:  GERING
undefined
Twaddle hydrometer
Catalog # GERI0549S
Supplier:  GERING

Specifications

  • Description:
    Twaddle hydrometer
  • Length:
    230 mm
  • Division:
    0,5 °TW
  • Pk:
    1
  • Range:
    48 – 74 °TW

Specifications

About this item

Glass Twaddle thermometer with calibration temperature of 20 °C.