Antibiotic assay discs
Supplier: PRAT DUMAS
Certificates
About this item
For determining antibiotic suited to the treatment of an infection and for checking their sensitivity to antibiotics by means of the inhibition zone determination method.
- Very thick cotton fibre papers, for the absorption of test solutions in antibiotic assays
- 420 g/m² weight
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together