Order Entry
Austria
ContactUsLinkComponent
Deep well plates, 96-well, PP, MegaBlock®
Deep well plates, 96-well, PP, MegaBlock®
Catalog # SARS82.1971.002
Supplier:  SARSTEDT
undefined
Deep well plates, 96-well, PP, MegaBlock®
Catalog # SARS82.1971.002
Supplier:  SARSTEDT

Specifications

  • Description:
    Deep well plate, 96-well, round wells, PS, 1,2 ml
  • Pk:
    32

Specifications

About this item

PP. The 96-well, 1,2/2,2 ml MegaBlock® meets all important requirements for automated sample processing or storing large quantities of backup samples.

  • Each well 100% leakproof tested
  • Suitable for DNA isolation, enzyme assays and tissue culture applications
  • Ideal for longterm storage of pharmaceutical samples
  • Solvent resistant to DMSO
  • Round bottom wells
  • Alphanumeric coding for easy identification