Order Entry
Austria
ContactUsLinkComponent
Precast horizontal gel kits, SERVALYT™ PRECOTES™ Tf Kit
Precast horizontal gel kits, SERVALYT™ PRECOTES™ Tf Kit
Catalog # SERA42803.01
Supplier:  Serva
Precast horizontal gel kits, SERVALYT™ PRECOTES™ Tf Kit
Catalog # SERA42803.01
Supplier:  Serva

Specifications

  • Description:
    SERVALYT™ PRECOTES™ TF kit, 245×125 mm
  • Seq:
    04
  • Pk:
    1

Specifications

About this item

For transferrin (Tf) analysis by isoelectric focusing.

SERVALYT™ PRECOTES™ Tf Kit contains:
5× SERVALYT™ PRECOTES™ Tf gels, 245×125 mm, 300 µm
Anode and cathode buffer solutions
Applicator strips and electrode wicks