Thermal desorber tubes
Supplier: PerkinElmer
Certificates
About this item
These desorber tubes are offered with a variety of sorbent packing materials for many GC applications including indoor and outdoor air monitoring, analysis of flavors and fragrances and the analysis of outgassing from packaging, polymers, pharmaceuticals and semi conductor material.
- Each tube maintains its unique serial number which is etched for easy identification
- With plastic end caps for short-term storage
Frequently Bought Together