Order Entry
Austria
Orders LinkContactUsLinkComponent
Cuvette rack

Cuvette rack

Cuvette rack, 16 (2×8), 210×70×35 mm
Catalog # HELL3001.10
Supplier:   Hellma
Cuvette rack
Catalog # HELL3001.10
Supplier:  Hellma
Cuvette rack

Cuvette rack, 16 (2×8), 210×70×35 mm

Price
20,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Polypropylene.

Rack for 16 square cells with 10 mm path length with inclined profile to aid cell handling.

Specifications
  • For:
    16 (2×8)
  • Description:
    Cuvette rack
  • W×D×H:
    210×70×35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.