Ice flakers, MF series
: SCOTSMAN
Modular ice flakers is designed to produce hygienic, clear, pure ice ideal for medical and laboratory applications. It requires both a water and electricity supply and air or water-cooled condenser. Stainless steel finish, with ABS plastic lid.
- Reduced-maintenance
- Self-lubricating gear-reducer
- Oversized hermetic compressor
- Removable side panels on all four sides to aid routine maintenance and servicing
: All units are delivered without a plug.
Frequently Bought Together