Bulk packing material for preparative HPLC columns, SepTech™ C18 reverse phase media
Supplier: VARIAN
Certificates
About this item
SepTech™ C18 bulk media, designed for high performance separations at high capacity. The two pore sizes, 60 Å and 150 Å, have been optimised to give high capacity for small molecules and larger natural molecules and peptides, respectively.
Frequently Bought Together