ContactUsLinkComponent
600 series Microliter™ syringes
600 series Microliter™ syringes
Supplier:  HAMILTON BONADUZ
Certificates
undefined
600 series Microliter™ syringes
Supplier:  HAMILTON BONADUZ
Specifications
  • Catalog No:
  • HAMI7633-01
  • HAMI7632-01
  • Fluid path:
  • Stainless steel, borosilicate glass
  • Stainless steel, borosilicate glass
  • Max. pressure:
  • 137,9 bar
  • 137,9 bar
  • Max. temperature:
  • 115 °C
  • 115 °C
  • Min. temperature:
  • 10 °C
  • 10 °C