Orders LinkContactUsLinkComponent
Electrode Plug Cable Combinations
Electrode Plug Cable Combinations
Supplier:   SI Analytics
Electrode Plug Cable Combinations
Supplier:  SI Analytics
Certificates
Electrode Plug Cable Combinations
Electrode Plug Cable Combinations
3 Options Available Below
Product Details & Documents
Electrode cables along with electrode socket, 1 m two core cable up to 80 °C, for resistance thermometer and 2-pole-conductivity cell without temperature sensor. Available with different cable length.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • SCOI285124613
    Plug cable combinations, electrode socket, 1 m two core cable up to 80 °C, free end
    Plug cable combinations, electrode socket, 1 m two core cable up to 80 °C, free end
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,90 €
    46,90 €
    1 * 1 ST
       46,90 €
  • SCOI285124654
    Plug cable combinations, electrode socket, 5 m two core cable up to 80 °C, free end
    Plug cable combinations, electrode socket, 5 m two core cable up to 80 °C, free end
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,00 €
    68,00 €
    1 * 1 ST
       68,00 €
  • SCOI285124708
    Plug cable combinations, electrode socket, 10 m two core cable up to 80 °C, free end
    Plug cable combinations, electrode socket, 10 m two core cable up to 80 °C, free end
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,00 €
    92,00 €
    1 * 1 ST
       92,00 €