Order Entry
Austria
Orders LinkContactUsLinkComponent
Eyewash station, QuickSafe® Basic

Eyewash station, QuickSafe® Basic

Eyewash station with cover, QuickSafe® Basic, with 2×500 ml Plum Eye Wash and 1×200 ml pH Neutral
Catalog # 118-0398
Supplier:   Plum Safety
Eyewash station, QuickSafe® Basic
Catalog # 118-0398
Supplier:  Plum Safety
Eyewash station, QuickSafe® Basic
Eyewash station, QuickSafe® Basic

Eyewash station with cover, QuickSafe® Basic, with 2×500 ml Plum Eye Wash and 1×200 ml pH Neutral

Price
112,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This wall-mountable station is made of impact-resistant plastic and equipped with a transparent cover.

  • Users can tailor the first shelf with products that fit their business
  • Cabinet is only slightly larger than an A3 sheet of paper
  • Comes with instructions and fixtures for wall mounting

Content:

2×500 ml Plum Eye Wash
1×200 ml pH Neutral
5×QuickRinse eyewash ampoules (20 ml)
1×QuickFix Elastic plaster refill

W×D×H: 253×92×430 mm

Specifications
  • Description:
    Eyewash station with cover, QuickSafe® Basic, with 2×500 ml Plum Eye Wash and 1×200 ml pH Neutral
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.