ContactUsLinkComponent
Wipe rolls, Multitex® DR
Wipe rolls, Multitex® DR
Catalog # 115-2743
undefined
Wipe rolls, Multitex® DR
Catalog # 115-2743
Specifications
  • Packed:
    8 rolls of 60 sheets
  • Colour:
    White
  • Description:
    Cleaning cloth Multitex® DR, roll for Wiper Bowl® Safe & Clean dispenser bucket
  • L×W:
    300×320 mm
  • Environmentally Preferable:
  • Pk:
    8 Roll
Specifications
About this item
These reusable wipes made from viscose fleece are suitable for disinfecting and cleaning applications. They are intended to be used with the Wiper Bowl® Safe and Clean dispenser bucket.
  • Perforated roll
  • Tear-resistant
  • Highly absorbent and lint-free