Orders LinkContactUsLinkComponent
RapID™ System
RapID™ System
RapID™ System
Certificates
RapID™ System
14 Options Available Below
Product Details & Documents
RapID™ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapID™ panels offer easy, one-step, simultaneous inoculation of dehydrated reactants in tray wells to identify anaerobes, coryneform Gram-positive bacilli, enterics, nonfermenters, streptococci, and yeast all within 4 hours. Systems are ideal for manual testing or as confirmation for automated test systems.
  • Rapid, accurate identification of over 400 medically important organisms
  • Simple, one-step inoculation
  • Four-hour aerobic incubation
  • Easy-to-read colour reactions
Documents
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • RapID™ One panel
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 213,00 €
    213,00 €
    1 * 20 ST
       213,00 €
  • OXOIR8325102
    RapID™ Inoculation Fluid 1 ml
    RapID™ Inoculation Fluid 1 ml
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 35,70 €
    35,70 €
    1 * 20 ST
       35,70 €
  • OXOIR8325106
    RapID™ Inoculation Fluid 2 ml
    RapID™ Inoculation Fluid 2 ml
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 39,10 €
    39,10 €
    1 * 20 ST
       39,10 €
  • OXOIR8311008
    RapID™ CB Plus panel
    RapID™ CB Plus panel
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 387,00 €
    387,00 €
    1 * 20 ST
       387,00 €
  • OXOIR8309002
    RapID™ Spot Indole reagent
    RapID™ Spot Indole reagent
    15 mL
    Log in for availability
    Log in for availability
    1 * 15 mL
    1 * 15 mL - 39,20 €
    39,20 €
    1 * 15 mL
       39,20 €
  • OXOIR8309003
    RapID™ Nitrate A reagent
    RapID™ Nitrate A reagent
    15 mL
    Log in for availability
    Log in for availability
    1 * 15 mL
    1 * 15 mL - 34,70 €
    34,70 €
    1 * 15 mL
       34,70 €
  • OXOIR8309004
    RapID™ Nitrate B reagent
    RapID™ Nitrate B reagent
    15 mL
    Log in for availability
    Log in for availability
    1 * 15 mL
    1 * 15 mL - 33,20 €
    33,20 €
    1 * 15 mL
       33,20 €
  • OXOIR8311001
    RapID™ NH panel
    RapID™ NH panel
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 252,00 €
    252,00 €
    1 * 20 ST
       252,00 €
  • OXOIR8311002
    RapID™ ANAII panel
    RapID™ ANAII panel
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 440,00 €
    440,00 €
    1 * 20 ST
       440,00 €
  • OXOIR8311003
    RapID™ Str panel
    RapID™ Str panel
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 418,00 €
    418,00 €
    1 * 20 ST
       418,00 €
  • OXOIR8311004
    RapID™ Ss/U panel
    RapID™ Ss/U panel
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 196,00 €
    196,00 €
    1 * 20 ST
       196,00 €
  • OXOIR8311007
    RapID™ Yeast Plus panel
    RapID™ Yeast Plus panel
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 380,00 €
    380,00 €
    1 * 20 ST
       380,00 €
  • OXOIR8323600
    RapID™ Eric
    RapID™ Eric
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,80 €
    34,80 €
    1 * 1 ST
       34,80 €
  • OXOIR8311009
    RapID™ Staph Plus System
    RapID™ Staph Plus System
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 506,00 €
    506,00 €
    1 * 20 ST
       506,00 €