Culture media, ready to use, in tubes
Supplier: MERCK LIFE SCIENCE
Certificates
About this item
The current editions of the United States Pharmacopoeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP) include harmonized methods for the microbiological examination of non-sterile pharmaceutical products. These methods address the total aerobic count (TAMC and TYMC) as well as the test for specified micro-organisms. Harmonized methods facilitate and accelerate the release of pharmaceutical products.
- A complete range of media required for testing all aerobic microorganisms in non-sterile pharmaceutical products
- The composition of all media is in accordance with the requirements of the harmonized chapters of the Pharmacopoeia
Frequently Bought Together