Culture media, ready to use, in tubes
Supplier: Dr. Möller & Schmelz
Certificates
About this item
These sterile ready-to-use media are filled in autoclavable and break-proof glass tubes (20 ml). The composition of the various types of media, comply with the formulations specified in the pertinent standards and regulations. Whereas the liquid media can be used immediately, the agar media first have to be remelted in a water bath at 90 to 100 °C. Once they have cooled down to around 50 °C they can be poured as agar plates in different sizes and thicknesses to meet individual demands.
- Delivered 50 ml in polycarbonate bottles or 20 ml in glass tubes
Frequently Bought Together