Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spinetch® D
Spinetch® D, VLSI Selectipur® for the electronics industry
Spinetch® D, VLSI Selectipur® for the electronics industry
Spinetch® D, VLSI Selectipur® for the electronics industry
Catalog # 50438060.
Supplier:
BASF ELECTRONIC CHEMICALS
CAS Number:
Certificates
Expand image
Spinetch® D, VLSI Selectipur® for the electronics industry
Catalog # 50438060.
Supplier:
BASF ELECTRONIC CHEMICALS
CAS Number:
Certificates
Specifications
Pack type:
Metal drum, lined
Pk:
276 kg
Seq:
4776
Specifications
About this item
Product Details
Documentation
:
EINECS:
0
Product Details
Documentation