Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fume Hoods and Enclosures
Chemical Fume Hoods
Accessories for laboratory hoods, Fiberglass 30
Accessories for laboratory hoods, Fiberglass 30
Accessories for laboratory hoods, Fiberglass 30
LBCP4882806
:
LABCONCO
Accessories for laboratory hoods, Fiberglass 30
LBCP4882806
:
LABCONCO
Description:
Solid epoxy work surface, flat, no cupsink cutout, for 30 inch width laboratory hoods, 760×760×30 mm, 39 kg
Pk:
1